Learn More
Medchemexpress LLC HY-119674A 100mg Medchemexpress, Xanthopterin (hydrate) CAS:5979-01-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY119674A100MG
Encompass_Preferred
Medchemexpress, HY-119674A 100mg Xanthopterin (hydrate) CAS:5979-01-1 Xanthopterin hydrate, an unconjugated pteridine compound, is the main component of the yellow granule in the Oriental hornet bear wings, produces a characteristic excitation/emission maximum at 386/456 nm. Xanthopterin hydrate(XPT) causes renal growth and hypertrophy in rat.Xanthopterin hydrate inhibits RNA synthesis[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9542