Learn More
AdipoGen Xanthorrhizol (1 mg)
SDP
Supplier:  AdipoGen AGCN20090M001
Encompass_Preferred
Chemical. CAS: 30199-26-9. Formula: C15H22O. MW: 218.3. Isolated from Curcuma xanthorrhiza. Shows calcium-antagonistic and vasorelaxant activity. Antibacterial. Anti-inflammatory. Potent COX-2 and inducible nitric oxide (iNOS; NOS II) inhibitor. Hepatoprotective. Anticancer compound. Apoptosis inducer. Neuroprotective antioxidant. Antifungal.
Catalog No. 50-205-3448