Learn More
Abcam x anthorrhizol, Antio x idant agent, 1MG
Catalog No. 502705818
Encompass_Preferred
MW 218.33 Da, Purity >97%. Antioxidant agent (IC₅₀ = 2 μM, copper-mediated oxidation of LDL). Antiproliferative agent. Activates caspase-3 and caspase-9. Modulates PARP-1 protein. Induces cytochrome c release. Shows apoptotic, antibacterial, antifungal, anticancer, phytoestrogenic, neuroprotective, antihyperglycemic and anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5818 Supplier Abcam Supplier No. AB1443871MG
MW 218.33 Da, Purity >97%. Antioxidant agent (IC₅₀ = 2 μM, copper-mediated oxidation of LDL). Antiproliferative agent. Activates caspase-3 and caspase-9. Modulates PARP-1 protein. Induces cytochrome c release. Shows apoptotic, antibacterial, antifungal, anticancer, phytoestrogenic, neuroprotective, antihyperglycemic and anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement