Learn More
Medchemexpress LLC Xanthosine 5'-monophosphate sodium salt | 25899-70-1 | 99.6% | 408.17 | 100 MG
SDP
Catalog No. 5000454050
Encompass_Preferred
Xanthosine 5'-monophosphate sodium salt is an intermediate in purine metabolism. It is a solid, white to light yellow in color, and soluble in H2O (100 mg/mL, 245.00 mM; requires ultrasonic).
  • Can be used for genetic code, nucleic acid structure, and DNA, RNA and protein synthesis research.
  • Potential use in foods as a flavoring agent.
  • Chemical intermediate for the production of purine antimetabolites.
  • Valuable in the production of synthetic polynucleotides for studies on the genetic code, protein synthesis, and nucleic acid structure.
Catalog No. 50-004-54050 Supplier Medchemexpress LLC Supplier No. HYW008990100MG
Xanthosine 5'-monophosphate sodium salt is an intermediate in purine metabolism. It is a solid, white to light yellow in color, and soluble in H2O (100 mg/mL, 245.00 mM; requires ultrasonic).
  • Can be used for genetic code, nucleic acid structure, and DNA, RNA and protein synthesis research.
  • Potential use in foods as a flavoring agent.
  • Chemical intermediate for the production of purine antimetabolites.
  • Valuable in the production of synthetic polynucleotides for studies on the genetic code, protein synthesis, and nucleic acid structure.