Learn More
Medchemexpress LLC HY-W011527 50mg Medchemexpress, Xanthosine CAS:146-80-5 Purity:>98%
SDP
Catalog No. 501934551
Encompass_Preferred
Medchemexpress, HY-W011527 50mg Xanthosine CAS:146-80-5 Xanthosine is a nucleoside derived from xanthine and ribose. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4551 Supplier Medchemexpress LLC Supplier No. HYW01152750MG
Medchemexpress, HY-W011527 50mg Xanthosine CAS:146-80-5 Xanthosine is a nucleoside derived from xanthine and ribose. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.