Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthosine Dihydrate 98.0+%, TCI America™
SDP

Catalog No. X0008100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0008100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X0008100MG Supplier TCI America Supplier No. X0008100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146-80-5
Molecular Formula C10H12N4O6
Molecular Weight (g/mol) 284.228
MDL Number MFCD00005726
InChI Key UBORTCNDUKBEOP-UUOKFMHZSA-N
PubChem CID 64959
ChEBI CHEBI:18107
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 284.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Xanthosine Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.