Learn More
Medchemexpress LLC HY-N1063 100mg Medchemexpress, Xanthoxylin CAS:90-24-4 Purity:>98%
SDP
Catalog No. 501970318
Encompass_Preferred
Medchemexpress, HY-N1063 100mg Xanthoxylin CAS:90-24-4 Xanthoxylin (Xanthoxyline) is isolated from Zanthoxylum simulans. Xanthoxylin (Xanthoxyline) has antifungal and antispasmodic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0318 Supplier Medchemexpress LLC Supplier No. HYN1063100MG
Medchemexpress, HY-N1063 100mg Xanthoxylin CAS:90-24-4 Xanthoxylin (Xanthoxyline) is isolated from Zanthoxylum simulans. Xanthoxylin (Xanthoxyline) has antifungal and antispasmodic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.