Learn More
American Radiolabeled Chemicals Inc XANTHURENIC AC 1LC
SDP
Catalog No. 50901192
Encompass_Preferred
Xanthurenic acid-ring-5,7-3H 1mCi in ethanol, 1 mCi/ml, S.A. 40-60 Ci/mmol, M.W. 205.17, Shipped in Dry Ice
Catalog No. 50-901-192 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 17601MC
Xanthurenic acid-ring-5,7-3H 1mCi in ethanol, 1 mCi/ml, S.A. 40-60 Ci/mmol, M.W. 205.17, Shipped in Dry Ice