Learn More
Medchemexpress LLC Xanthurenic acid | 59-00-7 | MFCD00006754 | 99.4% | 205.17 g/mol | C10H7NO4 | 50 MG
SDP
Catalog No. 5000302131
Encompass_Preferred
Xanthurenic acid (CAS 59-00-7) is a high-purity analytical reference standard for assay development, calibration, and compound identification. Supplied as a solid (off-white to light yellow), it is intended for use in chromatography, mass spectrometry, and other analytical workflows that require a well-characterized standard.
  • High purity reference standard (99.44%).
  • Suitable for chromatography and mass spectrometry.
  • Supplied as a solid, off-white to light yellow.
  • Available in multiple small pack sizes for analytical use.
  • Includes certificate of analysis and safety data sheet.
Catalog No. 50-003-02131 Supplier Medchemexpress LLC Supplier No. HYW014666R50MG
Add to Cart
Add to Cart
Xanthurenic acid (CAS 59-00-7) is a high-purity analytical reference standard for assay development, calibration, and compound identification. Supplied as a solid (off-white to light yellow), it is intended for use in chromatography, mass spectrometry, and other analytical workflows that require a well-characterized standard.
  • High purity reference standard (99.44%).
  • Suitable for chromatography and mass spectrometry.
  • Supplied as a solid, off-white to light yellow.
  • Available in multiple small pack sizes for analytical use.
  • Includes certificate of analysis and safety data sheet.