Learn More
Medchemexpress LLC Xanthurenic acid | 59-00-7 | MFCD28011017 | 99.4% | 205.17 g/mol | C10H7NO4 | 25 MG
SDP
Catalog No. 5000303404
Encompass_Preferred
Xanthurenic acid (CAS 59-00-7) is an analytical reference standard used in biochemical research and analytical testing. Provided as a solid (off-white to light yellow) with high reported purity, it is suitable for chromatography calibration, method development, and biochemical assays where a reliable endogenous metabolite standard is required.
  • High purity for analytical applications.
  • Solid form suitable for storage and handling.
  • Useful as a chromatography and calibration standard.
  • Suitable for method development and validation.
  • Relevant endogenous metabolite for biochemical studies.
Catalog No. 50-003-03404 Supplier Medchemexpress LLC Supplier No. 25MGHYW014666R
Add to Cart
Add to Cart
Xanthurenic acid (CAS 59-00-7) is an analytical reference standard used in biochemical research and analytical testing. Provided as a solid (off-white to light yellow) with high reported purity, it is suitable for chromatography calibration, method development, and biochemical assays where a reliable endogenous metabolite standard is required.
  • High purity for analytical applications.
  • Solid form suitable for storage and handling.
  • Useful as a chromatography and calibration standard.
  • Suitable for method development and validation.
  • Relevant endogenous metabolite for biochemical studies.