Learn More
Medchemexpress LLC Xanthurenic acid-d4 | 1329611-28-0 | MFCD27967185 | 99.1% | 209.19 | C10H3D4NO4 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HYW014666S2.5MG
Encompass_Preferred
Xanthurenic acid-d4 is the deuterium-labeled analogue of xanthurenic acid supplied as a solid for research use. It is formulated for use as an internal standard in analytical workflows, providing consistent isotopic shifts for accurate quantitation in mass spectrometry and chromatographic assays.
  • Deuterium-labeled for isotope-dilution and mass spectrometry quantitation.
  • High chemical purity suitable for analytical standards.
  • Molecular weight 209.19 g/mol and formula C10H3D4NO4 provide exact mass reference.
  • Compatible with LC-MS/MS and GC-MS workflows.
  • Available in small, accurately weighed quantities for calibration and method validation.
  • Supplied as a light yellow to yellow solid; intended for research use only.
Catalog No. 50-003-02642