Learn More
Strem, An Ascensus Company CAS# 161265-03-8. 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, min. 98% XANTPHOS. MFCD00233866. 100G
SDP
Supplier:  Strem, An Ascensus Company 151242100G
Encompass
CAS# 161265-03-8. 100G. 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, min. 98% XANTPHOS. MFCD00233866. Molecular Formula: C39H32OP2. Molecular Weight: 578.63. Color/Form: light-yellow xtl. Strem#: 15-1242 www.strem.com/catalog/v3/15-1242/
Catalog No. 50-195-9523