Learn More
Strem, An Ascensus Company XantphosPalladacycleG4, 98% 5g
SDP
Supplier:  Strem, An Ascensus Company 4603885G
Encompass
CAS# 1621274-19-8. 5g. Methanesulfonato[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene](2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), 98% [Xantphos Palladacycle Gen. 4]. MFCD28411658. Molecular Weight: 962.38. Molecular Formula: C53H47NO4P2PdS. Color/form: yellow solid Strem# 46-0388. http://www.strem.com/catalog/v/46-0388/
Catalog No. 50-203-4171