Learn More
Medchemexpress LLC HY-15147 10mg , XAV-939 CAS:284028-89-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1514710MG
Encompass_Preferred
HY-15147 10mg XAV-939 CAS: 284028-89-3 XAV-939 is a potent tankyrase inhibitor that targets Wnt/β-catenin signaling. XAV-939 stabilizes axin by inhibiting tankyrase 1 and tankyrase 2 (IC50s of 5 and 2 nM, respectively), thereby stimulating β-catenin degradation. XAV939 binds tightly to the catalytic (PARP) domains of TNKS1 and TNKS2 (Kds of 99 and 93 nM, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9146