Learn More
Selleck Chemical LLC XAV-939 10mM/1mL 284028-89-3 NVP-XAV939
SDP
Catalog No. 501360990
Encompass_Preferred
XAV-939 (NVP-XAV939) selectively inhibits Wnt/?-catenin-mediated transcription through tankyrase1/2 inhibition with IC50 of 11 nM/4 nM in cell-free assays, regulates axin levels and does not affect CRE, NF-?B or TGF-?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0990 Supplier Selleck Chemical LLC Supplier No. S118010MM/1ML
XAV-939 (NVP-XAV939) selectively inhibits Wnt/?-catenin-mediated transcription through tankyrase1/2 inhibition with IC50 of 11 nM/4 nM in cell-free assays, regulates axin levels and does not affect CRE, NF-?B or TGF-?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.