Learn More
Apexbio Technology LLC XAV-939, 5mg. Cas: 284028-89-3 MFCD: MFCD00106866. Tankyrase 1/2 inhibitor
SDP
Supplier:  Apexbio Technology LLC A18775
Encompass_Preferred
XAV-939 is a small-molecule inhibitor of tankyrase 1/2 with IC50 values of 4 and 11 nM, respectively [1].XAV-939 was screened out as an inhibitor of Wnt/-catenin pathway. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4714