Learn More
Medchemexpress LLC XAV-939 (GMP) | 284028-89-3 | C14H11F3N2OS | 50 MG
SDP
Catalog No. 5000372196
Encompass_Preferred
XAV-939 (GMP) is a tankyrase inhibitor produced using GMP guidelines. It functions as an auxiliary reagent for cell therapy manufacture and induces human pluripotent stem cells (hPSCs) to differentiate into post-mitotic cortical neurons, promoting anterior CNS identity.
  • Functions as an auxiliary reagent for cell therapy manufacture
  • Induces human pluripotent stem cells (hPSCs) to post-mitotic cortical neurons differentiation
  • Promotes anterior CNS identity
  • Tankyrase inhibitor
  • Produced using GMP guidelines
Catalog No. 50-003-72196 Supplier Medchemexpress LLC Supplier No. HY15147G50MG
XAV-939 (GMP) is a tankyrase inhibitor produced using GMP guidelines. It functions as an auxiliary reagent for cell therapy manufacture and induces human pluripotent stem cells (hPSCs) to differentiate into post-mitotic cortical neurons, promoting anterior CNS identity.
  • Functions as an auxiliary reagent for cell therapy manufacture
  • Induces human pluripotent stem cells (hPSCs) to post-mitotic cortical neurons differentiation
  • Promotes anterior CNS identity
  • Tankyrase inhibitor
  • Produced using GMP guidelines