Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XAV 939 98.0+%, TCI America™
SDP

Catalog No. X0077100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0077100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X0077100MG Supplier TCI America Supplier No. X0077100MG

Chemical Identifiers

CAS 284028-89-3
Molecular Formula C14H11F3N2OS
Molecular Weight (g/mol) 312.31
MDL Number MFCD16879017
InChI Key KLGQSVMIPOVQAX-UHFFFAOYSA-N
Synonym 3,5,7,8-tetrahydro-2-4-trifluoromethyl phenyl-4h-thiopyrano 4,3-d pyrimidin-4-one, 2-4-trifluoromethyl phenyl-7,8-dihydro-5h-thiopyrano 4,3-d pyrimidin-4-ol, 2-4-trifluoromethyl phenyl-7,8-dihydro-5h-thiino 4,3-d pyrimidin-4-ol, 2-4-trifluoromethyl phenyl-5h,7h,8h-thiopyrano 4,3-d pyrimidin-4-ol, 2-4-trifluoromethyl phenyl-1,5,7,8-tetrahydrothiopyrano 4,3-d pyrimidin-4-one, 2-4-trifluoromethyl phenyl-1,5,7,8-tetrahydro-4h-thiopyrano 4,3-d pyrimidin-4-one, 2-4-trifluoromethyl phenyl-1h,5h,7h,8h-thiopyrano 4,3-d pyrimidin-4-one, 2-4-trifluoromethyl phenyl-3h,5h,7h,8h-thiopyrano 4,3-d pyrimidin-4-one
PubChem CID 2726824
ChEBI CHEBI:62878
IUPAC Name 2-[4-(trifluoromethyl)phenyl]-1,5,7,8-tetrahydrothiopyrano[4,3-d]pyrimidin-4-one
SMILES C1CSCC2=C1NC(=NC2=O)C3=CC=C(C=C3)C(F)(F)F

Specifications

Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 312.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material XAV 939
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.