Learn More
Enzo Life Sciences XAV939 (5 mg). CAS: 284028-89-3
SDP
Catalog No. 502012357
Encompass_Preferred
Potent inhibitor of Tankyrase 1 (IC50=11nM) and Tankyrase 2 (IC50=4nM). Inhibition of tankyrase promotes stabilization of axin resulting in β-catenin degradation and Wnt signalling disruption. Inhibits proliferation of the μ-catenin-dependent colon carcinoma cell line DLD-1. Promotes cardiomyogenic development in mesoderm progenitor cells. Alternative name: 3,5,7,8-Tetrahydro-2-[4-(trifluoromethyl)phenyl]-4H-thiopyrano[4,3-d]pyrimidin-4-one. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-2357 Supplier Enzo Life Sciences Supplier No. BMLWN1000005
Potent inhibitor of Tankyrase 1 (IC50=11nM) and Tankyrase 2 (IC50=4nM). Inhibition of tankyrase promotes stabilization of axin resulting in β-catenin degradation and Wnt signalling disruption. Inhibits proliferation of the μ-catenin-dependent colon carcinoma cell line DLD-1. Promotes cardiomyogenic development in mesoderm progenitor cells. Alternative name: 3,5,7,8-Tetrahydro-2-[4-(trifluoromethyl)phenyl]-4H-thiopyrano[4,3-d]pyrimidin-4-one. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20 mg/ml). Long Term Storage: Ambient.