Learn More
Abcam x AV939, Tankyrase inhibitor, 5MG
Catalog No. 502707151
Encompass_Preferred
MW 312.31 g/mol, Purity >98%. Potent tankyrase (TNKS) inhibitor. Inhibitor of Wnt/β-catenin signaling. (IC₅₀ values are 11 and 4 nM for TNKS1 and 2, respectively). Inhibits TNKS PARP activity. Selectivity over other PARP enzymes and CRE, NK-κB and TGF-β.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7151 Supplier Abcam Supplier No. AB1208975MG
MW 312.31 g/mol, Purity >98%. Potent tankyrase (TNKS) inhibitor. Inhibitor of Wnt/β-catenin signaling. (IC₅₀ values are 11 and 4 nM for TNKS1 and 2, respectively). Inhibits TNKS PARP activity. Selectivity over other PARP enzymes and CRE, NK-κB and TGF-β.
The product is subject to the following: Abcam Restricted Use Statement