Learn More
WELCH MATERIALS INC Ultisil XB-C18, 40-70µm, 100Å, 1kg
Catalog No. NC2052745
Encompass_Preferred
Bulk Media, Ultisil XB-C18, 40-70μm, pH range: 1.5-10.0, Pore Size: 100Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped. Packing: 1kg/bottle.
Catalog No. NC2052745 Supplier WELCH MATERIALS INC Supplier No. H0271011600
Bulk Media, Ultisil XB-C18, 40-70μm, pH range: 1.5-10.0, Pore Size: 100Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped. Packing: 1kg/bottle.