Learn More
WELCH MATERIALS INC Ultisil XB-C18, 5µm, 120Å, 7.8×250mm
Catalog No. NC2052188
Encompass_Preferred
Analytical Column, Ultisil XB-C18, 5μm, 120Å, 7.8×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052188 Supplier WELCH MATERIALS INC Supplier No. H0020131051
Analytical Column, Ultisil XB-C18, 5μm, 120Å, 7.8×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped.