Learn More
WELCH MATERIALS INC Ultisil XB-C8, 10µm, 120Å, 4.6×250mm
Catalog No. NC2052209
Encompass_Preferred
Analytical Column, Ultisil XB-C8, 10μm, 120Å, 4.6×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L7, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052209 Supplier WELCH MATERIALS INC Supplier No. H0020241043
Analytical Column, Ultisil XB-C8, 10μm, 120Å, 4.6×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L7, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.