Learn More
WELCH MATERIALS INC Ultisil XB-NH2, 10µm, 30×250mm
Catalog No. NC2052709
Encompass_Preferred
Prep Column, Ultisil XB-NH2, 10µm, 30×250mm, pH range: 2.0-8.0, Pore Size: 120Å, USP listing: L8, Carbon Loading: 4%, Surface Area: 320m2/g, not-endcapped.
Catalog No. NC2052709 Supplier WELCH MATERIALS INC Supplier No. H0263031108
Add to Cart
Add to Cart
Request a Quote
Prep Column, Ultisil XB-NH2, 10µm, 30×250mm, pH range: 2.0-8.0, Pore Size: 120Å, USP listing: L8, Carbon Loading: 4%, Surface Area: 320m2/g, not-endcapped.