Learn More
WELCH MATERIALS INC Ultisil XB-NH2, 5µm, 120Å, 7.8×250mm
Catalog No. NC2052220
Encompass_Preferred
Analytical Column, Ultisil XB-NH2, 5μm, 120Å, 7.8×250mm, pH range: 2.0-8.0, Pore Size: 120Å, USP listing: L8, Carbon Loading: 4%, Surface Area: 320m2/g, not-endcapped.
Catalog No. NC2052220 Supplier WELCH MATERIALS INC Supplier No. H0020431051
Analytical Column, Ultisil XB-NH2, 5μm, 120Å, 7.8×250mm, pH range: 2.0-8.0, Pore Size: 120Å, USP listing: L8, Carbon Loading: 4%, Surface Area: 320m2/g, not-endcapped.