Learn More
New England Biolabs, Inc. XbaI – 15000 units
SDP
Catalog No. 50812021
Encompass
A restriction endonuclease that recognizes the sequence T^CTAG_A.
Catalog No. 50-812-021 Supplier New England Biolabs, Inc. Supplier No. R0145M
A restriction endonuclease that recognizes the sequence T^CTAG_A.