Learn More
New England Biolabs, Inc. XbaI – 15000 units
SDP
Catalog No. 50812020
Encompass
A restriction endonuclease that recognizes the sequence T^CTAG_A.
Catalog No. 50-812-020 Supplier New England Biolabs, Inc. Supplier No. R0145L
A restriction endonuclease that recognizes the sequence T^CTAG_A.