Learn More
New England Biolabs, Inc. XbaI – 3000 units
SDP
Catalog No. 50812022
Encompass
A restriction endonuclease that recognizes the sequence T^CTAG_A.
Catalog No. 50-812-022 Supplier New England Biolabs, Inc. Supplier No. R0145S
A restriction endonuclease that recognizes the sequence T^CTAG_A.