Learn More
Thermo Scientific™ XBF40 Wireless Triple Timer
Catalog No. 13990414 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
XBF40D-MD -40°C blast freezer
Catalog No. For Use With (Equipment)
13-990-414 XBF40D-MD -40°C blast freezer
1 options
Catalog No. 13-990-414 Supplier Thermo Scientific™ Supplier No. 6642
Add to Cart
Add to Cart
Wireless, triple timer tracks the freezing at each shelf separately.
Keep track of the rapid freezing of plasma in the XBF40D-MD -40°C blast freezer. The Thermo Scientific™ XBF40 Wireless Triple Timer tracks each shelf separately.
J316
TRUSTED_SUSTAINABILITY
Specifications
Description XBF40 Wireless Triple Timer
Type Wireless Triple Timer
Temperature Range -40°C
For Use With (Equipment) XBF40D-MD -40°C blast freezer