Learn More
Enzo Life Sciences XBP 1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2230100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human XBP-1, spliced isoform XBP1(S). Source: Purified from rabbit serum. Species reactivity: Human, Mouse. Applications: Flow Cytometry, ICC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-2772