Learn More
Enzo Life Sciences XBP-1S monoclonal antibody (8F6.11) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2550100
Encompass_Preferred
Clone: 8F6.11. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human XBP-1, spliced isoform XBP1(S). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Applications: ELISA, ICC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-2800