Learn More
Aviva Systems Biology Corp XBP1 Antibody - N-terminal region (ARP38552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human XBP1. Alternate names: TREB5, XBP2ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-56700