Learn More
Aviva Systems Biology Corp XBP1 Antibody - C-terminal region (ARP38553_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38553_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human XBP1. Alternate names: TREB5, XBP2ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56701