Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Glycan BEH Amide Column, 130Å, 3.5 μm, 4.6 mm × 150 mm

Catalog No. 50727015 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column uses BEH-based Glycan chemistry, supporting UPLC and HPLC with scalable particle sizes, including 3.5 μm. Each batch is QC tested with a 2-AB labeled Glycan Performance standard for consistent glycan analysis.

  • Optimized for Hydrophilic Interaction (HILIC) separation mode.
  • Constructed with a hybrid particle substrate.
  • Operates effectively within a pH range of 2 to 11.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Features low silanol activity.
  • Suitable for molecular weights ranging from 1,000 to 150,000.
  • Composed of spherical particles with a size of 3.5 μm.
  • Has a pore size of 130 Å.
  • Quality control tested for glycan applications.
  • Designed for HPLC systems.
  • Inner diameter of 4.6 mm.
  • Length of 150 mm.

Catalog No. 50-727-015 Supplier Waters Corp Supplier No. 186007275
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This column uses BEH-based Glycan chemistry, supporting UPLC and HPLC with scalable particle sizes, including 3.5 μm. Each batch is QC tested with a 2-AB labeled Glycan Performance standard for consistent glycan analysis.

  • Optimized for Hydrophilic Interaction (HILIC) separation mode.
  • Constructed with a hybrid particle substrate.
  • Operates effectively within a pH range of 2 to 11.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Features low silanol activity.
  • Suitable for molecular weights ranging from 1,000 to 150,000.
  • Composed of spherical particles with a size of 3.5 μm.
  • Has a pore size of 130 Å.
  • Quality control tested for glycan applications.
  • Designed for HPLC systems.
  • Inner diameter of 4.6 mm.
  • Length of 150 mm.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.