Learn More
Waters Corp Waters XBridge BEH Amide Column, 130 Å, 5 μm, 2.1 mm x 150 mm
Catalog No. 50466343 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Amide columns are designed to retain extremely polar compounds, including carbohydrates and sugars. These columns feature a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11, facilitating the separation of polar analytes with varying polarity, structural moiety, and pKa.
  • Retains extremely polar compounds, such as carbohydrates and sugars.
  • Features a chemically stable, trifunctionally bonded amide phase.
  • Stable across a pH range of 2 to 11.
  • Enables separation of polar analytes with varying polarity, structural moiety, and pKa.
Catalog No. 50-466-343 Supplier Waters Corp Supplier No. 186006590
XBridge BEH Amide columns are designed to retain extremely polar compounds, including carbohydrates and sugars. These columns feature a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11, facilitating the separation of polar analytes with varying polarity, structural moiety, and pKa.
  • Retains extremely polar compounds, such as carbohydrates and sugars.
  • Features a chemically stable, trifunctionally bonded amide phase.
  • Stable across a pH range of 2 to 11.
  • Enables separation of polar analytes with varying polarity, structural moiety, and pKa.