Learn More
Waters Corp Waters Column, Amide, 5 μm, 2.1 mm x 30 mm
Catalog No. 50466340 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These BEH Amide columns are designed to retain extremely polar compounds, such as carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase that maintains stability across a broad pH range of 2 to 11, facilitating the separation of polar analytes with diverse polarity, structural characteristics, and pKa values.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Enables separation of polar analytes spanning a wide range of polarity, structural moiety, and pKa.
Catalog No. 50-466-340 Supplier Waters Corp Supplier No. 186006587
These BEH Amide columns are designed to retain extremely polar compounds, such as carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase that maintains stability across a broad pH range of 2 to 11, facilitating the separation of polar analytes with diverse polarity, structural characteristics, and pKa values.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Enables separation of polar analytes spanning a wide range of polarity, structural moiety, and pKa.