Learn More
Waters Corp Column, Amide Chemistry, 5 μm, 130 Å, 2.1 mm x 50 mm
Catalog No. 50466341 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Amide Column is designed to retain extremely polar compounds like carbohydrates and sugars. It features a chemically stable, trifunctionally bonded amide phase, stable across a pH range of 2 to 11, facilitating HILIC separations. Utilizing ethylene bridged hybrid (BEH) particle technology, it offers superior stability for carbohydrate analysis, high-resolution, and rapid analysis even in challenging sample matrices. It has a particle size of 5 μm and a pore size of 130 Å.
  • Retains extremely polar compounds (carbohydrates and sugars).
  • Chemically stable amide phase, stable from pH 2 to 11.
  • Superior stability and adaptability for HILIC separations.
  • Novel ethylene bridged hybrid (BEH) particle technology for extended column lifetimes and performance.
Catalog No. 50-466-341 Supplier Waters Corp Supplier No. 186006588
The XBridge BEH Amide Column is designed to retain extremely polar compounds like carbohydrates and sugars. It features a chemically stable, trifunctionally bonded amide phase, stable across a pH range of 2 to 11, facilitating HILIC separations. Utilizing ethylene bridged hybrid (BEH) particle technology, it offers superior stability for carbohydrate analysis, high-resolution, and rapid analysis even in challenging sample matrices. It has a particle size of 5 μm and a pore size of 130 Å.
  • Retains extremely polar compounds (carbohydrates and sugars).
  • Chemically stable amide phase, stable from pH 2 to 11.
  • Superior stability and adaptability for HILIC separations.
  • Novel ethylene bridged hybrid (BEH) particle technology for extended column lifetimes and performance.