Learn More
Waters Corp Waters XBridge BEH Amide Column, 5 μm, 3 mm x 100 mm
Catalog No. 50466345 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is an XBridge BEH Amide column designed for separating polar analytes. It offers chemical stability across a wide pH range (2 to 11), making it suitable for retaining highly polar compounds like carbohydrates and sugars. Its trifunctionally bonded amide phase ensures robust and reliable performance for various applications.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Offers a chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11, enabling separation of polar analytes across a wide range of polarity, structural moiety, and pKa.
  • Features low silanol activity.
  • Utilizes spherical particle technology.
Catalog No. 50-466-345 Supplier Waters Corp Supplier No. 186006592
Add to Cart
Add to Cart
This product is an XBridge BEH Amide column designed for separating polar analytes. It offers chemical stability across a wide pH range (2 to 11), making it suitable for retaining highly polar compounds like carbohydrates and sugars. Its trifunctionally bonded amide phase ensures robust and reliable performance for various applications.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Offers a chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11, enabling separation of polar analytes across a wide range of polarity, structural moiety, and pKa.
  • Features low silanol activity.
  • Utilizes spherical particle technology.