Learn More
Waters Corp Waters Column, Amide, 5 μm, 4.6 mm, 250 mm
Supplier:  Waters Corp 186006596
Encompass_Preferred
These columns are designed to retain extremely polar compounds like carbohydrates and sugars. They feature a chemically stable amide phase, stable from pH 2 to 11, facilitating separation of polar analytes. They provide increased mass spectrometry response and are compatible with sample preparation eluates.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Enables separation of polar analytes across a wide range of polarity.
  • Offers increased mass spectrometry response.
  • Direct compatibility with sample preparation eluates.
  • Orthogonal selectivity compared to reversed-phase materials.
  • Manufactured using strict processes for reproducibility.
Catalog No. 50-466-349