Learn More
Waters Corp Waters XBridge BEH Amide Column, 130Å, 3.5 μm, 2.1 mm X 50 mm
Catalog No. 50791213 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Amide columns are for HILIC separations, retaining polar compounds like carbohydrates. They feature a chemically stable amide phase, stable from pH 2–11, and utilize BEH particle technology for high resolution.
  • Retains polar compounds.
  • Chemically stable amide phase.
  • pH 2–11 stable.
  • BEH particle technology.
  • HILIC stability and versatility.
  • Seamless HPLC/UPLC transfer.
  • Wide mobile phase pH support.
  • 3.5 μm particle, 130 Å pore for resolution.
Catalog No. 50-791-213 Supplier Waters Corp Supplier No. 186004859
XBridge BEH Amide columns are for HILIC separations, retaining polar compounds like carbohydrates. They feature a chemically stable amide phase, stable from pH 2–11, and utilize BEH particle technology for high resolution.
  • Retains polar compounds.
  • Chemically stable amide phase.
  • pH 2–11 stable.
  • BEH particle technology.
  • HILIC stability and versatility.
  • Seamless HPLC/UPLC transfer.
  • Wide mobile phase pH support.
  • 3.5 μm particle, 130 Å pore for resolution.