Learn More
Waters Corp Waters Amide Column, 3.5 μm, 1 mm x 50 mm
Catalog No. 50466120 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Amide columns are designed to retain extremely polar compounds like carbohydrates and sugars. These columns feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11. They enable separation of polar analytes across a wide range of polarity and pKa, using a novel ethylene bridged hybrid (BEH) particle for superior stability in HILIC separations.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11.
  • Separates polar analytes with varying polarity and pKa.
  • Based on novel ethylene bridged hybrid (BEH) particle.
  • Allows transfer of separations between HPLC and UPLC technology platforms.
  • Ideal for carbohydrate analysis, enabling high resolution in challenging matrices.
  • Manufactured under strict quality assurance standards.
Catalog No. 50-466-120 Supplier Waters Corp Supplier No. 186004871
Add to Cart
Add to Cart
XBridge BEH Amide columns are designed to retain extremely polar compounds like carbohydrates and sugars. These columns feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11. They enable separation of polar analytes across a wide range of polarity and pKa, using a novel ethylene bridged hybrid (BEH) particle for superior stability in HILIC separations.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11.
  • Separates polar analytes with varying polarity and pKa.
  • Based on novel ethylene bridged hybrid (BEH) particle.
  • Allows transfer of separations between HPLC and UPLC technology platforms.
  • Ideal for carbohydrate analysis, enabling high resolution in challenging matrices.
  • Manufactured under strict quality assurance standards.