Learn More
Waters Corp Waters XBridge BEH Amide Column, 130Å, 2.5 μm, 2.1 mm x 150 mm, 3-pack
Supplier: Waters Corp 176002889
XBridge BEH Amide columns are designed to retain extremely polar compounds, including carbohydrates and sugars. They feature a chemically stable amide phase, stable across a broad pH range of 2 to 11, facilitating the separation of diverse polar analytes. Packed with 2.5 μm eXtended Performance (XP) ultra-low dispersion hardware, these columns deliver exceptional separation performance and high throughput for HPLC assays, providing a smooth pathway for UPLC integration.
- Stable across a wide pH range, from 2 to 11.
- Ultra-low dispersion hardware for eXtended Performance (XP).
- Provides exceptional separation performance, robustness, and high throughput for HPLC assays.
- Seamless transition path toward UPLC adoption.
- Chemistry: Amide.
- Separation mode: Hydrophilic Interaction (HILIC).
- Particle size: 2.5 μm.
- Pore size: 130 Å.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.