Learn More
Waters Corp XBridge BEH Amide Method Validation Kit, 130Å, 2.5 µm, 2.1 mm X 150 mm, 3/pk
Catalog No. 50466474 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three Xbridge BEH Amide columns packed with three different batches of sorbent. XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars. XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-474 Supplier Waters Corp Supplier No. 186006782
This method validation kit consists of three Xbridge BEH Amide columns packed with three different batches of sorbent. XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars. XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.