Learn More
Waters Corp Waters Corporation XBridge BEH Amide Method Validation Kit, 2.5 μm, 130 Å, 4.6 mm x 150 mm
Catalog No. 50466476 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit features three XBridge BEH Amide columns, designed for retaining extremely polar compounds like carbohydrates. They utilize a chemically stable amide phase, stable from pH 2 to 11, for broad analyte separation. Packed in ultra-low dispersion hardware, 2.5 μm XP columns ensure high performance for HPLC, with a clear path to UPLC.
  • Amide chemistry; HILIC separation mode
  • Hybrid particle substrate
  • pH stable from 2 to 11
  • Maximum pressure: 10000 psi (690 Bar)
  • Spherical particle shape, low silanol activity
  • BEH particle technology for stability
  • Includes 3 columns per package
Catalog No. 50-466-476 Supplier Waters Corp Supplier No. 186006784
This method validation kit features three XBridge BEH Amide columns, designed for retaining extremely polar compounds like carbohydrates. They utilize a chemically stable amide phase, stable from pH 2 to 11, for broad analyte separation. Packed in ultra-low dispersion hardware, 2.5 μm XP columns ensure high performance for HPLC, with a clear path to UPLC.
  • Amide chemistry; HILIC separation mode
  • Hybrid particle substrate
  • pH stable from 2 to 11
  • Maximum pressure: 10000 psi (690 Bar)
  • Spherical particle shape, low silanol activity
  • BEH particle technology for stability
  • Includes 3 columns per package