Learn More
Waters Corp Waters XBridge Column, BEH Amide, 2.5 μm, 3 mm x 150 mm
Supplier:  Waters Corp 186006725
Encompass_Preferred
XBridge BEH Amide columns are designed to retain extremely polar compounds, such as carbohydrates and sugars, featuring a chemically stable, trifunctionally bonded amide phase stable from pH 2 to 11. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, providing exceptional separation performance, robustness, and throughput for HPLC assays, and enabling a smooth transition to UPLC.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable amide phase, stable from pH 2 to 11.
  • Ultra-low dispersion hardware for exceptional performance in HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • Offers orthogonal selectivity compared to reversed-phase materials.
  • Exceptional column lifetimes due to BEH particle technology.
Catalog No. 50-466-433