Learn More
Waters Corp Waters Column, Amide, 2.5 μm, 130 Å, 4.6 mm, 150 mm
Catalog No. 50466434 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Amide columns are designed for the retention of extremely polar compounds such as carbohydrates and sugars. They feature a chemically stable amide phase, stable from pH 2 to 11, facilitating the separation of polar analytes. These 2.5 μm XP columns deliver exceptional performance for HPLC assays and enable a seamless transition to UPLC, resisting pressures up to 10,000 psi (690 bar).
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Ultra-low dispersion hardware for exceptional separation performance, robustness, and throughput.
  • Enables seamless transition path toward UPLC adoption.
  • Resistant to pressures up to 10,000 psi (690 bar).
  • Provides remarkable column longevity due to proprietary bonding techniques and end-capping.
Catalog No. 50-466-434 Supplier Waters Corp Supplier No. 186006726
XBridge BEH Amide columns are designed for the retention of extremely polar compounds such as carbohydrates and sugars. They feature a chemically stable amide phase, stable from pH 2 to 11, facilitating the separation of polar analytes. These 2.5 μm XP columns deliver exceptional performance for HPLC assays and enable a seamless transition to UPLC, resisting pressures up to 10,000 psi (690 bar).
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Ultra-low dispersion hardware for exceptional separation performance, robustness, and throughput.
  • Enables seamless transition path toward UPLC adoption.
  • Resistant to pressures up to 10,000 psi (690 bar).
  • Provides remarkable column longevity due to proprietary bonding techniques and end-capping.