Learn More
Waters Corp XBridge BEH Amide Column, 130 Å, 5 μm, 4.6 mm, 250 mm
Catalog No. 502392305 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Amide columns offer robust separation of polar compounds in HPLC applications.
  • Designed to retain extremely polar compounds (e.g., carbohydrates, sugars).
  • Chemically stable, trifunctionally bonded amide phase, pH 2–11 stable.
  • Electronic eConnect column tag for auto-identification, tracking, and digital tracing.
  • Compatible with any HPLC system; optimized for Alliance iS System with Empower software.
  • Hydrophilic Interaction (HILIC) separation mode.
  • Hybrid particle substrate with low silanol activity and spherical shape.
  • Particle size: 5 μm; Pore size: 130 Å; Max pressure: 6000 psi (415 Bar).
  • BEH particle technology; USP classification L68; Small molecule application.
  • Units per package: 1.
Catalog No. 50-239-2305 Supplier Waters Corp Supplier No. 186006596RF
XBridge BEH Amide columns offer robust separation of polar compounds in HPLC applications.
  • Designed to retain extremely polar compounds (e.g., carbohydrates, sugars).
  • Chemically stable, trifunctionally bonded amide phase, pH 2–11 stable.
  • Electronic eConnect column tag for auto-identification, tracking, and digital tracing.
  • Compatible with any HPLC system; optimized for Alliance iS System with Empower software.
  • Hydrophilic Interaction (HILIC) separation mode.
  • Hybrid particle substrate with low silanol activity and spherical shape.
  • Particle size: 5 μm; Pore size: 130 Å; Max pressure: 6000 psi (415 Bar).
  • BEH particle technology; USP classification L68; Small molecule application.
  • Units per package: 1.