Learn More
Waters Corp Waters Corporation Chromatography Column, XBridge BEH Amide, 3.5 μm, 130 Å, 2.1 mm X 100 mm
Supplier:  Waters Corp 186004860
Encompass_Preferred
XBridge BEH Amide columns retain extremely polar compounds like carbohydrates and sugars. They use a chemically stable, trifunctionally bonded amide phase, stable across pH 2 to 11, for diverse polar analyte separations. Built on BEH particle technology, these columns offer stability and versatility in HILIC, optimized for high-resolution carbohydrate analysis with 3.5 μm particle size and 130Å pore size.
  • Chemically stable, trifunctionally bonded amide phase ensures stability in HILIC.
  • Wide pH compatibility (2 to 11) for diverse polar analyte retention.
  • Exceptional retention for polar analytes across broad polarity and pKa.
  • Optimized for high-resolution carbohydrate analysis (3.5 μm, 130Å).
  • Seamless separation transfer between HPLC and UPLC platforms.
Catalog No. 50-203-7682