Learn More
Waters Corp Waters XBridge BEH Amide Column, 130Å, 3.5 μm, 3 mm × 100 mm
Catalog No. 502037683 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Amide columns are designed to retain extremely polar compounds, including carbohydrates and sugars. These columns provide a chemically stable, trifunctionally bonded amide phase, stable from pH 2–11, allowing for the separation of polar analytes with varying polarity, structural moiety, and pKa.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Offers a chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2–11, enabling the separation of polar analytes spanning a wide range of polarity, structural moiety, and pKa.
Catalog No. 50-203-7683 Supplier Waters Corp Supplier No. 186004864
XBridge BEH Amide columns are designed to retain extremely polar compounds, including carbohydrates and sugars. These columns provide a chemically stable, trifunctionally bonded amide phase, stable from pH 2–11, allowing for the separation of polar analytes with varying polarity, structural moiety, and pKa.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Offers a chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2–11, enabling the separation of polar analytes spanning a wide range of polarity, structural moiety, and pKa.