Learn More
Waters Corp XBridge BEH Amide OBD Prep Column, 130Å, 5 μm, 10 mm x 50 mm
Catalog No. 502391017 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Amide OBD Prep Column, with 130Å pore, 5 μm particle, and 10 mm x 50 mm dimensions, is designed for preparative HPLC. It separates polar analytes with exceptional resistance to mechanical bed failure, offering consistent, scalable performance from analytical to preparative dimensions.
  • Enables fast, efficient lab-scale separations for greater throughput.
  • Offers higher recoveries and extended column lifetimes.
  • Provides exceptional resistance to mechanical bed failure.
  • Delivers consistent column-to-column performance.
  • Assures scalability by matching analytical and preparative column bed densities.
  • Maximizes productivity when scaling screening methods to preparative HPLC.
  • Offers diverse BEH Technology chemistries for ruggedness and longevity at low and high pH.
Catalog No. 50-239-1017 Supplier Waters Corp Supplier No. 186008260
The XBridge BEH Amide OBD Prep Column, with 130Å pore, 5 μm particle, and 10 mm x 50 mm dimensions, is designed for preparative HPLC. It separates polar analytes with exceptional resistance to mechanical bed failure, offering consistent, scalable performance from analytical to preparative dimensions.
  • Enables fast, efficient lab-scale separations for greater throughput.
  • Offers higher recoveries and extended column lifetimes.
  • Provides exceptional resistance to mechanical bed failure.
  • Delivers consistent column-to-column performance.
  • Assures scalability by matching analytical and preparative column bed densities.
  • Maximizes productivity when scaling screening methods to preparative HPLC.
  • Offers diverse BEH Technology chemistries for ruggedness and longevity at low and high pH.